3-Isocyan-4-chlorbenzotrifluorid

Lieferant: CHEMPUR

FL21208.POR
CHMPFL21208.POREA 0 EUR
CHMPFL21208.POR
3-Isocyan-4-chlorbenzotrifluorid
3-Isocyan-4-chlorbenzotrifluorid
Formel: C₈H₃ClF₃N
CAS-Nummer: 601465-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR