4-Isocyan-3-nitrobenzotrifluorid

Lieferant: CHEMPUR

FL21209.POR
CHMPFL21209.POREA 0 EUR
CHMPFL21209.POR
4-Isocyan-3-nitrobenzotrifluorid
4-Isocyan-3-nitrobenzotrifluorid
Formel: C₈H₃F₃N₂O₂
MDL: MFCD04117597
CAS-Nummer: 1029634-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR