5-Isocyan-2-chlorbenzotrifluorid

Lieferant: CHEMPUR

FL21210.POR
CHMPFL21210.POREA 0 EUR
CHMPFL21210.POR
5-Isocyan-2-chlorbenzotrifluorid
5-Isocyan-2-chlorbenzotrifluorid
Formel: C₈H₃ClF₃N
CAS-Nummer: 220145-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR