2,6-Xylylisocyanid

Lieferant: CHEMPUR

FL21215.POR
CHMPFL21215.POREA 0 EUR
CHMPFL21215.POR
2,6-Xylylisocyanid
2,6-Xylylisocyanid
Formel: C₉H₉N
MDL: MFCD00013479
CAS-Nummer: 2769-71-3
EINECS: 220-459-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR