2,3-Dimethylphenylisocyanat
Lieferant: CHEMPUR
Synonyme:
2,3-Xylylisocyanat
FL21216.POR
CHMPFL21216.POREA
0
EUR
CHMPFL21216.POR
2,3-Dimethylphenylisocyanat
2,3-Dimethylphenylisocyanat
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Siedepunkt: 104…105 °C (50 mmHg) Dichte: 1,059 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00013851 CAS-Nummer: 1591-99-7 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...