2-Isocyan-5-chlorbenzotrifluorid

Lieferant: CHEMPUR

FL21230.POR
CHMPFL21230.POREA 0 EUR
CHMPFL21230.POR
2-Isocyan-5-chlorbenzotrifluorid
2-Isocyan-5-chlorbenzotrifluorid
Formel: C₈H₃ClF₃N
CAS-Nummer: 2920-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR