3-Acetyl-2,5-dimethylthiophen

Lieferant: CHEMPUR

FL2781.25
CHMPFL2781.25EA 0 EUR
CHMPFL2781.25
3-Acetyl-2,5-dimethylthiophen
3-Acetyl-2,5-dimethylthiophen
Formel: C₈H₁₀OS
Molecular Weight: 154,23 g/mol
MDL: MFCD00009763
CAS-Nummer: 2530-10-1
EINECS: 219-779-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR