3-(Methylsulfanyl)butanal

Lieferant: FLUOROCHEM

Synonyme: 3-(Methylthio)butyraldehyde

002806-5G FL2806.25 002806-1G
FLUO002806-5GEA 0 EUR
FLUO002806-5G CHMPFL2806.25 FLUO002806-1G
3-(Methylsulfanyl)butanal
3-(Methylthio)butanal
Formel: C₅H₁₀OS
Molecular Weight: 118.2 g/mol
MDL: MFCD00036622
CAS-Nummer: 16630-52-7
EINECS: 240-678-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR