(2-Chlorethyl)phenylsulfon

Lieferant: CHEMPUR

Synonyme: [(2-Chlorethyl)sulfonyl]benzol

FL3121.POR
CHMPFL3121.POREA 0 EUR
CHMPFL3121.POR
(2-Chlorethyl)phenylsulfon
(2-Chlorethyl)phenylsulfon
Formel: C₈H₉ClO₂S
MDL: MFCD00025047
CAS-Nummer: 938-09-0
EINECS: 213-337-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR