2-Fluorstyrol

Lieferant: CHEMPUR

Synonyme: 1-Fluoro-2-vinylbenzene

FL3236.25
CHMPFL3236.25EA 0 EUR
CHMPFL3236.25
2-Fluorstyrol
2-Fluorstyrol
Formel: C₈H₇F
Molecular Weight: 122,14 g/mol
Siedepunkt: 94 °C (0,5 mmHg)
Dichte: 1,03 g/cm³ (20 °C)
MDL: MFCD00013550
CAS-Nummer: 394-46-7
EINECS: 206-896-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR