3,3'-Methylendianilin

Lieferant: CHEMPUR

Synonyme: 3,3'-Methylenedianiline, 3,3'-Diaminodiphenylmethane

FL3375.POR
CHMPFL3375.POREA 0 EUR
CHMPFL3375.POR
3,3'-Methylendianilin
3,3'-Methylendianilin
Formel: C₁₃H₁₄N₂
MDL: MFCD00014777
CAS-Nummer: 19471-12-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR