3,4'-Methylendianilin

Lieferant: CHEMPUR

Synonyme: 3,4'-Diaminodiphenylmethane, 3,4'-Methylenedianiline

FL3376.POR
CHMPFL3376.POREA 0 EUR
CHMPFL3376.POR
3,4'-Methylendianilin
3,4'-Methylendianilin
Formel: C₁₃H₁₄N₂
MDL: MFCD00014778
CAS-Nummer: 19430-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR