6-Methylnicotinsäure

Lieferant: CHEMPUR

Synonyme: 6-Methyl-3-pyridincarbonsäure, 6-Methylpyridin-3-carbonsäure

FL3382.500 003382-1G 003382-5G 003382-25G
CHMPFL3382.500EA 0 EUR
CHMPFL3382.500 FLUO003382-1G FLUO003382-5G FLUO003382-25G
6-Methylnicotinsäure
6-Methylnicotinsäure
Formel: C₇H₇NO₂
Schmelzpunkt: 212 °C
MDL: MFCD00006341
CAS-Nummer: 3222-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR