6-Phenyl-1-hexanol

Lieferant: CHEMPUR

Synonyme: 6-Phenylhexanol

FL3386.1
CHMPFL3386.1EA 0 EUR
CHMPFL3386.1
6-Phenyl-1-hexanol
6-Phenyl-1-hexanol
Formel: C₁₂H₁₈O
Molecular Weight: 178,27 g/mol
MDL: MFCD00014063
CAS-Nummer: 2430-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR