2-Bromethyl-ethylsulfid

Lieferant: CHEMPUR

FL3467.POR
CHMPFL3467.POREA 0 EUR
CHMPFL3467.POR
2-Bromethyl-ethylsulfid
2-Bromethyl-ethylsulfid
Formel: C₄H₉BrS
MDL: MFCD00236595
CAS-Nummer: 35420-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR