Diethyl-aminomalonat Hydrochlorid
Lieferant: CHEMPUR
Synonyme:
Diethyl 2-aminomalonate hydrochloride
FL3624.POR
CHMPFL3624.POREA
0
EUR
CHMPFL3624.POR
Diethyl-aminomalonat Hydrochlorid
Diethyl-aminomalonat Hydrochlorid
Formel:
C₇H₁₄ClNO₄ Molecular Weight: 211,65 g/mol |
MDL:
MFCD00012510 CAS-Nummer: 13433-00-6 EINECS: 236-556-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...