3-Amino-2-(2-fluorphenoxy)pyridin

Lieferant: CHEMPUR

FL3989.POR
CHMPFL3989.POREA 0 EUR
CHMPFL3989.POR
3-Amino-2-(2-fluorphenoxy)pyridin
3-Amino-2-(2-fluorophenoxy)pyridine
Formel: C₁₁H₉FN₂O
MDL: MFCD00067806
CAS-Nummer: 175135-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR