4'-Brom-3'-(trifluormethyl)acetanilid

Lieferant: CHEMPUR

Synonyme: N-(4-Bromo-3-(trifluoromethyl)phenyl)acetamide

CFF430.25 CFF430.50 FL4304.5
CHMPCFF430.25EA 481 EUR
CHMPCFF430.25 CHMPCFF430.50 CHMPFL4304.5
4'-Brom-3'-(trifluormethyl)acetanilid
4'-Brom-3'-(trifluormethyl)acetanilid
Formel: C₉H₇BrF₃NO
Molecular Weight: 282,07 g/mol
MDL: MFCD00052361
CAS-Nummer: 41513-05-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR