ω-Chlor-ω,ω-difluoracetophenon

Lieferant: CHEMPUR

Synonyme: 2-Chlor-2,2-difluoracetophenon, α-Chlor-α,α-difluoracetophenon

FL4392.1
CHMPFL4392.1EA 0 EUR
CHMPFL4392.1
ω-Chlor-ω,ω-difluoracetophenon
ω-Chlor-ω,ω-difluoracetophenon
Formel: (C₆H₅)C(O)CClF₂
MDL: MFCD00042123
CAS-Nummer: 384-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR