1,1-Dibromdifluorethylen

Lieferant: CHEMPUR

FL4644.POR
CHMPFL4644.POREA 0 EUR
CHMPFL4644.POR
1,1-Dibromdifluorethylen
1,1-Dibromdifluorethylen
Formel: C₂Br₂F₂
CAS-Nummer: 430-85-3
EINECS: 207-068-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR