4-Fluor-2,6-dinitrophenol

Lieferant: CHEMPUR

FL4968.1
CHMPFL4968.1EA 0 EUR
CHMPFL4968.1
4-Fluor-2,6-dinitrophenol
4-Fluor-2,6-dinitrophenol
Formel: C₆H₃FN₂O₅
Molecular Weight: 202,1 g/mol
MDL: MFCD00179333
CAS-Nummer: 364-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR