2-Fluorthiophen

Lieferant: CHEMPUR

FL5314.500 FL5314.1
CHMPFL5314.500EA 0 EUR
CHMPFL5314.500 CHMPFL5314.1
2-Fluorthiophen
2-Fluorothiophene
Formel: C₄H₃FS
MDL: MFCD02181179
CAS-Nummer: 400-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR