4-Butoxybenzaldehyd

Lieferant: CHEMPUR
Achtung

FL5341.POR 005341-25G 005341-100G
CHMPFL5341.POREA 0 EUR
CHMPFL5341.POR FLUO005341-25G FLUO005341-100G
4-Butoxybenzaldehyd
4-Butoxybenzaldehyd
Formel: C₁₁H₁₄O₂
Molecular Weight: 178.23 g/mol
Siedepunkt: 285
Dichte: 1.0285 g/cm³ (20 °C)
MDL: MFCD00003389
CAS-Nummer: 5736-88-9
EINECS: 227-247-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR