3,4,5-Trifluor-o-phenylendiamin

Lieferant: CHEMPUR

Synonyme: 1,2-Diamino-3,4,5-trifluorbenzol, 3,4,5-Trifluorbenzol-1,2-diamin, 3,4,5-Trifluor-1,2-phenylendiamin

FL5486.1
CHMPFL5486.1EA 0 EUR
CHMPFL5486.1
3,4,5-Trifluor-o-phenylendiamin
3,4,5-Trifluor-o-phenylendiamin
Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
MDL: MFCD01569524
CAS-Nummer: 168966-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR