2-Chlor-6-fluorzimtsäure

Lieferant: CHEMPUR

FL5635.1 FL5635.5
CHMPFL5635.1EA 0 EUR
CHMPFL5635.1 CHMPFL5635.5
2-Chlor-6-fluorzimtsäure
2-Chlor-6-fluorzimtsäure
Formel: C₉H₆ClFO₂
MDL: MFCD00051582
CAS-Nummer: 392-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR