2,3-Difluor-6-nitrophenol

Lieferant: CHEMPUR

Synonyme: 1,2-Difluoro-3-hydroxy-4-nitrobenzene

FL5776.POR
CHMPFL5776.POREA 0 EUR
CHMPFL5776.POR
2,3-Difluor-6-nitrophenol
2,3-Difluor-6-nitrophenol
Formel: C₆H₃F₂NO₃
Schmelzpunkt: 61 °C
MDL: MFCD00042255
CAS-Nummer: 82419-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR