2-Methyl-4-(trifluormethoxy)benzamid

Lieferant: CHEMPUR

FL5835.POR
CHMPFL5835.POREA 0 EUR
CHMPFL5835.POR
2-Methyl-4-(trifluormethoxy)benzamid
2-Methyl-4-(trifluormethoxy)benzamid
Formel: C₉H₈F₃NO₂
MDL: MFCD01631494
CAS-Nummer: 261951-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR