2,5-Dimethylresorcin

Lieferant: CHEMPUR

Synonyme: 2,5-Dimethylbenzene-1,3-diol

FL5920.POR
CHMPFL5920.POREA 0 EUR
CHMPFL5920.POR
2,5-Dimethylresorcin
2,5-Dimethylresorcin
Formel: C₈H₁₀O₂
Molecular Weight: 138,1638 g/mol
MDL: MFCD00134473
CAS-Nummer: 488-87-9
EINECS: 207-688-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR