1-(Bromdifluormethyl)cyclohexen

Lieferant: CHEMPUR

FL5926.POR
CHMPFL5926.POREA 0 EUR
CHMPFL5926.POR
1-(Bromdifluormethyl)cyclohexen
1-(Bromodifluoromethyl)cyclohexene
Formel: C₇H₉BrF₂
MDL: MFCD00153695
CAS-Nummer: 117711-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR