1-Fluor-2,4,6-trimethylpyridiniumtetrafluoroborat

Lieferant: CHEMPUR

Synonyme: 1-Fluoro-2,4,6-trimethylpyridinium Tetrafluoroborate

FL6456.POR
CHMPFL6456.POREA 0 EUR
CHMPFL6456.POR
1-Fluor-2,4,6-trimethylpyridiniumtetrafluoroborat
1-Fluoro-2,4,6-trimethylpyridiniumtetrafluoroborate
Formel: C₈H₁₁BF₅N
Molecular Weight: 226,98 g/mol
MDL: MFCD00153182
CAS-Nummer: 109705-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR