1-Fluor-2,4,6-trimethylpyridiniumtrifluormethansulfonat

Lieferant: CHEMPUR

Synonyme: 1-Fluoro-2,4,6-trimethylpyridin-1-ium trifluoromethanesulphonate

FL6457.POR
CHMPFL6457.POREA 0 EUR
CHMPFL6457.POR
1-Fluor-2,4,6-trimethylpyridiniumtrifluormethansulfonat
1-Fluoro-2,4,6-trimethylpyridiniumtrifluoromethanesulphonate
Formel: C₉H₁₁F₄NO₃S
MDL: MFCD00067525
CAS-Nummer: 107264-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR