4-Nitro-2-(trifluormethyl)anisol

Lieferant: CHEMPUR

Synonyme: 2-Methoxy-5-nitrobenzotrifluorid, 1-Methoxy-4-nitro-2-(trifluormethyl)benzol

FL6744.10
CHMPFL6744.10EA 0 EUR
CHMPFL6744.10
4-Nitro-2-(trifluormethyl)anisol
4-Nitro-2-(trifluormethyl)anisol
Formel: C₈H₆F₃NO₃
Molecular Weight: 221,14 g/mol
Schmelzpunkt: 79…79,5 °C
MDL: MFCD00007328
CAS-Nummer: 654-76-2
EINECS: 211-507-3
UN: 2811
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR