2-(Difluormethoxy)phenylessigsäure

Lieferant: CHEMPUR

FL7867.POR
CHMPFL7867.POREA 0 EUR
CHMPFL7867.POR
2-(Difluormethoxy)phenylessigsäure
2-(Difluormethoxy)phenylessigsäure
Formel: C₉H₈F₂O₃
MDL: MFCD00236237
CAS-Nummer: 86867-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR