3-(Difluormethoxy)phenylessigsäure

Lieferant: CHEMPUR

FL7868.POR
CHMPFL7868.POREA 0 EUR
CHMPFL7868.POR
3-(Difluormethoxy)phenylessigsäure
3-(Difluormethoxy)phenylessigsäure
Formel: C₉H₈F₂O₃
MDL: MFCD00236238
CAS-Nummer: 262587-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR