4-(Difluormethoxy)phenylessigsäure

Lieferant: CHEMPUR

Synonyme: 2-(4-(Difluoromethoxy)phenyl)acetic acid

FL7869.POR
CHMPFL7869.POREA 0 EUR
CHMPFL7869.POR
4-(Difluormethoxy)phenylessigsäure
4-(Difluormethoxy)phenylessigsäure
Formel: C₉H₈F₂O₃
Molecular Weight: 202,1600 g/mol
MDL: MFCD00236239
CAS-Nummer: 243659-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR