2'-Trifluormethoxyacetophenon
Lieferant: CHEMPUR
Synonyme:
1-[2-(Trifluormethoxy)phenyl]-1-ethanon
FL7976.POR
007976-1G
007976-5G
007976-25G
CHMPFL7976.POREA
0
EUR
CHMPFL7976.POR
FLUO007976-1G
FLUO007976-5G
FLUO007976-25G
2'-Trifluormethoxyacetophenon
2'-Trifluormethoxyacetophenon
Formel:
C₉H₇F₃O₂ Molecular Weight: 204.15 |
MDL:
MFCD00236320 CAS-Nummer: 220227-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...