3-(Trifluormethylthio)benzylbromid

Lieferant: CHEMPUR

FL8011.1 FL8011.5
CHMPFL8011.1EA 0 EUR
CHMPFL8011.1 CHMPFL8011.5
3-(Trifluormethylthio)benzylbromid
3-(Trifluormethylthio)benzylbromid
Formel: C₈H₆BrF₃S
MDL: MFCD00236346
CAS-Nummer: 213203-84-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR