(±)-2-Phenyl-3-butin-2-ol

Lieferant: CHEMPUR

Synonyme: 2-Phenylbut-3-yn-2-ol

FL8691.POR
CHMPFL8691.POREA 0 EUR
CHMPFL8691.POR
(±)-2-Phenyl-3-butin-2-ol
(±)-2-Phenyl-3-butin-2-ol
Formel: C₁₀H₁₀O
Molecular Weight: 146,19 g/mol
MDL: MFCD00004454
CAS-Nummer: 127-66-2
EINECS: 204-855-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR