2-Brom-5-fluoranilin

Lieferant: CHEMPUR
Achtung

FL8853.POR
CHMPFL8853.POREA 0 EUR
CHMPFL8853.POR
2-Brom-5-fluoranilin
2-Brom-5-fluoranilin
Formel: C₆H₅BrFN
Molecular Weight: 190,01 g/mol
MDL: MFCD00070750
CAS-Nummer: 1003-99-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR