2,4-Dichlorphenylhydraziniumchlorid

Lieferant: CHEMPUR

Synonyme: 1-(2,4-Dichlorphenyl)hydrazin Hydrochlorid, 2,4-Dichlorphenylhydrazinhydrochlorid, 2,4-Dichlorphenylhydrazin Hydrochlorid

CFF1011. FL8957.POR 008957-100G 008957-25G
CHMPCFF1011.EA 0 EUR
CHMPCFF1011. CHMPFL8957.POR FLUO008957-100G FLUO008957-25G
2,4-Dichlorphenylhydraziniumchlorid
2,4-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
Molecular Weight: 213.49 g/mol
MDL: MFCD00012929
CAS-Nummer: 5446-18-4
EINECS: 226-659-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR