4-Amino-3-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

009014-1G FL9014.5 009014-500MG
FLUO009014-1GEA 525 EUR
FLUO009014-1G CHMPFL9014.5 FLUO009014-500MG
4-Amino-3-(trifluormethyl)pyridin
4-Amino-3-(trifluoromethyl)pyridine
Formel: C₆H₅F₃N₂
Molecular Weight: 162.11 g/mol
MDL: MFCD02183554
CAS-Nummer: 387824-61-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR