1,5-Decadiin

Lieferant: CHEMPUR

FL9116.10
CHMPFL9116.10EA 0 EUR
CHMPFL9116.10
1,5-Decadiin
1,5-Decadiin
Formel: C₁₀H₁₄
Molecular Weight: 134.22 g/mol
MDL: MFCD00041651
CAS-Nummer: 53963-03-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR