2-Fluor-5-nitrobenzoesäure
Lieferant: CHEMPUR
|
Achtung
|
CFF1134.
FL9222.10
009222-25G
009222-5G
009222-100G
CHMPCFF1134.EA
0
EUR
CHMPCFF1134.
CHMPFL9222.10
FLUO009222-25G
FLUO009222-5G
FLUO009222-100G
2-Fluor-5-nitrobenzoesäure
2-Fluor-5-nitrobenzoesäure
Formel:
C₇H₄FNO₄ |
MDL:
MFCD00134238 CAS-Nummer: 7304-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...