1H,1H,2H,3H,3H-Tridecafluor-1,2-nonandiol

Lieferant: CHEMPUR

Synonyme: 1H,1H,2H,3H,3H-Perfluor-1,2-nonandiol, 4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluor-1,2-nonandiol

FL9475.POR
CHMPFL9475.POREA 0 EUR
CHMPFL9475.POR
1H,1H,2H,3H,3H-Tridecafluor-1,2-nonandiol
1H,1H,2H,3H,3H-Tridecafluor-1,2-nonandiol
Formel: C₉H₇F₁₃O₂
Molecular Weight: 394,13 g/mol
MDL: MFCD00042262
CAS-Nummer: 107650-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR