3-Methoxyphenylisothiocyanat

Lieferant: CHEMPUR

Synonyme: 3-Methoxyphenylisothiocyanate

FL9513.POR 009513-25G 009513-5G
CHMPFL9513.POREA 0 EUR
CHMPFL9513.POR FLUO009513-25G FLUO009513-5G
3-Methoxyphenylisothiocyanat
3-Methoxyphenylisothiocyanat
Formel: C₈H₇NOS
Molecular Weight: 165.22 g/mol
Siedepunkt: 84 °C (1 mmHg)
Dichte: 1.1788 g/cm³ (20 °C)
MDL: MFCD00040335
CAS-Nummer: 3125-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR