(2-Hydroxylpropyl)-β-cyclodextrin

Lieferant: CHEMPUR

FL9871.POR
CHMPFL9871.POREA 0 EUR
CHMPFL9871.POR
(2-Hydroxylpropyl)-β-cyclodextrin
(2-Hydroxylpropyl)-β-cyclodextrin
Formel: C₆₃H₁₁₂O₄₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16621721
CAS-Nummer: 128446-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR