4-Amino-3,5-dimethyl-1,2,4-triazol

Lieferant: CHEMPUR

FL9907.POR
CHMPFL9907.POREA 0 EUR
CHMPFL9907.POR
4-Amino-3,5-dimethyl-1,2,4-triazol
4-Amino-3,5-dimethyl-1,2,4-triazole
Formel: C₄H₈N₄
Molecular Weight: 112,14 g/mol
MDL: MFCD00051647
CAS-Nummer: 3530-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR