4-Amino-3,5-dipropyl-1,2,4-triazol

Lieferant: CHEMPUR

FL9908.POR 009908-1G 009908-5G
CHMPFL9908.POREA 0 EUR
CHMPFL9908.POR FLUO009908-1G FLUO009908-5G
4-Amino-3,5-dipropyl-1,2,4-triazol
4-Amino-3,5-dipropyl-1,2,4-triazole
Formel: C₈H₁₆N₄
MDL: MFCD00744864
CAS-Nummer: 46054-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR