Diethylcyclobutan-1,1-dicarboxylat

Lieferant: CHEMPUR

JFD00475.250 JFD00475.1
CHMPJFD00475.250EA 0 EUR
CHMPJFD00475.250 MAYBJFD00475.1
Diethylcyclobutan-1,1-dicarboxylat
Diethylcyclobutan-1,1-dicarboxylat
Formel: C₁₀H₁₆O₄
MDL: MFCD00019261
CAS-Nummer: 3779-29-1
EINECS: 223-239-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR