4-(2-Hydroxyethyl)phenol
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
Tyrosol, 4-Hydroxyphenethylalkohol, 2-(4-Hydroxyphenyl)ethylalkohol
KS-5274-10MG
CHMPKS-5274-10MGEA
0
EUR
CHMPKS-5274-10MG
4-(2-Hydroxyethyl)phenol
2-(4-Hydroxyphenyl)ethanol
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 90…92 °C |
MDL:
MFCD00002902 CAS-Nummer: 501-94-0 EINECS: 207-930-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...